Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoin 98%

Supplier: AMBEED, INC
Warning

A714786-1000G A714786-100G A714786-25G A714786-500G
76775-992EA 41 USD InStock
76775-992 76775-994 76775-996 76775-998
Benzoin 98%
Benzoin
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00004496
CAS Number: 119-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty