Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethoxycinnamic acid 98%

Supplier: AMBEED, INC
Warning

A715826-25G A715826-5G
76776-008EA 58 USD InStock
76776-008 76776-010
2,5-Dimethoxycinnamic acid 98%
2,5-Dimethoxycinnamic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 375
Melting Pt: 150
MDL Number: MFCD00004378
CAS Number: 10538-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty