Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4,4,4-trifluoro-3-hydroxybutyrate 96%

Supplier: AMBEED, INC
Danger

A719833-1G A719833-250MG A719833-25G A719833-5G
76776-044EA 10 USD InStock
76776-044 76776-046 76776-048 76776-050
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate 96%
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate
Precaución: for Research Use Only


Formula: C₆H₉F₃O₃
MDL Number: MFCD00040842
CAS Number: 372-30-5
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty