Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Acetylpicolinic acid 97%

Supplier: AMBEED, INC
Warning

A744204-100MG A744204-1G A744204-250MG
76776-180EA 67 USD InStock
76776-180 76776-182 76776-184
6-Acetylpicolinic acid 97%
6-Acetylpicolinic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₃
MW: 165.148 g/mol
MDL Number: MFCD09800364
CAS Number: 122637-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty