Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Phenylglycine ethyl ester hydrochloride 98%

Supplier: AMBEED, INC
Warning

A750350-100G A750350-25G A750350-5G
76776-222EA 117 USD InStock
76776-222 76776-224 76776-226
DL-Phenylglycine ethyl ester hydrochloride 98%
DL-Phenylglycine ethyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂·ClH
MW: 215.677 g/mol
MDL Number: MFCD00114583
CAS Number: 879-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty