Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-benzoxazolethiol 98%

Supplier: AMBEED, INC
Warning

A769187-100G A769187-1G A769187-25G A769187-5G
76776-342EA 218 USD InStock
76776-342 76776-344 76776-346 76776-348
6-Chloro-2-benzoxazolethiol 98%
6-Chloro-2-benzoxazolethiol
Precaución: for Research Use Only


Formula: C₇H₄ClNOS
MDL Number: MFCD00800495
CAS Number: 22876-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty