Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-2-quinolinol 98%

Supplier: AMBEED, INC
Warning

A801851-1G A801851-250MG A801851-5G
76776-520EA 48 USD InStock
76776-520 76776-522 76776-524
6-Methyl-2-quinolinol 98%
6-Methyl-2-quinolinol
Precaución: for Research Use Only


Formula: C₁₀H₉NO
MDL Number: MFCD08236767
CAS Number: 4053-34-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty