Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(trifluoromethyl)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A804280-1G A804280-25G A804280-10G
76776-546EA 7 USD InStock
76776-546 76776-548 77627-592
3-Fluoro-4-(trifluoromethyl)benzaldehyde 98%
3-Fluoro-4-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₄F₄O
MDL Number: MFCD00236275
CAS Number: 204339-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty