Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine 98%

Supplier: AMBEED, INC
Warning

A818413-1G A818413-250MG A818413-5G
76776-598EA 134 USD InStock
76776-598 76776-600 76776-602
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine 98%
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine
Precaución: for Research Use Only


Formula: C₆H₅ClN₄
MDL Number: MFCD00975127
CAS Number: 7197-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty