Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',4',5'-Trimethoxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A958200-100G A958200-1G A958200-25G A958200-5G
76777-038EA 279 USD InStock
76777-038 76777-040 76777-042 76777-044
3',4',5'-Trimethoxyacetophenone 98%
3',4',5'-Trimethoxyacetophenone
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₄
MDL Number: MFCD00008738
CAS Number: 1136-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty