Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-methyl-2H-indazole 98%

Supplier: AMBEED, INC
Warning

A101220-100MG A101220-1G A101220-250MG
76780-474EA 8 USD InStock
76780-474 76780-476 76780-478
6-Chloro-2-methyl-2H-indazole 98%
6-Chloro-2-methyl-2H-indazole
Precaución: for Research Use Only


Formula: C₈H₇ClN₂
MDL Number: MFCD11977536
CAS Number: 541539-87-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty