Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

methyl 4-cyano-3-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A102437-10G A102437-1G A102437-5G
76780-546EA 83 USD InStock
76780-546 76780-548 76780-550
methyl 4-cyano-3-methylbenzoate 98%
methyl 4-cyano-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MDL Number: MFCD11655969
CAS Number: 25978-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty