Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Tetrahydrofurfuryl alcohol 98%

Supplier: AMBEED, INC
Warning

A103169-1000G A103169-100G A103169-500G
76780-608EA 31 USD InStock
76780-608 76780-610 76780-612 76780-614
(±)-Tetrahydrofurfuryl alcohol 98%
(±)-Tetrahydrofurfuryl alcohol
Precaución: for Research Use Only


Formula: C₅H₁₀O₂
MDL Number: MFCD00005372
CAS Number: 97-99-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty