Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl orthoisobutyrate 95%

Supplier: AMBEED, INC
Danger

A103604-1G A103604-25G A103604-5G
76780-656EA 9 USD InStock
76780-656 76780-658 76780-660
Trimethyl orthoisobutyrate 95%
Trimethyl orthoisobutyrate
Precaución: for Research Use Only


Formula: C₇H₁₆O₃
MDL Number: MFCD08276428
CAS Number: 52698-46-1
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty