Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cbz-L-tyrosine benzyl ester 97%

Supplier: AMBEED, INC
Warning

A104250-100G A104250-10G A104250-1G A104250-250MG A104250-25G A104250-500G A104250-5G
76780-686EA 122 USD InStock
76780-686 76780-688 76780-690 76780-692 76780-694 76780-696 76780-698
Cbz-L-tyrosine benzyl ester 97%
Cbz-L-tyrosine benzyl ester
Precaución: for Research Use Only


Formula: C₂₄H₂₃NO₅
MDL Number: MFCD00136670
CAS Number: 5513-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty