Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tyrphostin AG-1296 99%

Supplier: AMBEED, INC
Warning

A104725-10MG A104725-1MG A104725-25MG A104725-5MG A104725-100MG A104725-250MG A104725-50MG
76780-746EA 41 USD InStock
76780-746 76780-748 76780-750 76780-752 77666-524 77666-526 77666-528
Tyrphostin AG-1296 99%
Tyrphostin AG-1296
Precaución: for Research Use Only


Formula: C₁₆H₁₄N₂O₂
MW: 266.3 g/mol
MDL Number: MFCD00270913
CAS Number: 146535-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty