Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-chloropyrimidin-2-amine 98%

Supplier: AMBEED, INC
Warning

A106418-100G A106418-1G A106418-250MG A106418-25G A106418-5G
76780-836EA 592 USD InStock
76780-836 76780-838 76780-840 76780-842 76780-844
5-Bromo-4-chloropyrimidin-2-amine 98%
5-Bromo-4-chloropyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₄H₃BrClN₃
MDL Number: MFCD11046979
CAS Number: 1044767-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty