Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-2-Nal-OH 97%

Supplier: AMBEED, INC
Warning

A107621-1G A107621-25G A107621-5G
76780-910EA 15 USD InStock
76780-910 76780-912 76780-914
Z-2-Nal-OH 97%
Z-2-Nal-OH
Precaución: for Research Use Only


Formula: C₂₁H₁₉NO₄
MDL Number: MFCD00063338
CAS Number: 65365-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty