Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Boc-2-(Hydroxymethyl)piperidine 97%

Supplier: AMBEED, INC
Warning

A108056-100MG A108056-10G A108056-1G A108056-250MG
76780-934EA 5 USD InStock
76780-934 76780-936 76780-938 76780-940
(S)-1-Boc-2-(Hydroxymethyl)piperidine 97%
(S)-1-Boc-2-(Hydroxymethyl)piperidine
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD04973121
CAS Number: 134441-93-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty