Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl isoquinoline-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A108949-10G A108949-250MG A108949-5G
76780-984EA 108 USD InStock
76780-984 76780-986 76780-988
Ethyl isoquinoline-3-carboxylate 98%
Ethyl isoquinoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MDL Number: MFCD10566077
CAS Number: 50458-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty