Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributylammonium pyrophosphate 98%

Supplier: AMBEED, INC
Warning

A118000-100MG A118000-1G A118000-250MG A118000-5G A118000-25G
76781-500EA 34 USD InStock
76781-500 76781-502 76781-504 76781-506 77549-368
Tributylammonium pyrophosphate 98%
Tributylammonium pyrophosphate
Precaución: for Research Use Only


Formula: C₂₄H₅₈N₂O₇P₂
MDL Number: MFCD00136019
CAS Number: 5975-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty