Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide 97%

Supplier: AMBEED, INC
Warning

A119665-10G A119665-1G A119665-250MG A119665-25G A119665-5G
76781-658EA 43 USD InStock
76781-658 76781-660 76781-662 76781-664 76781-666
Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide 97%
Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide
Precaución: for Research Use Only


Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
MDL Number: MFCD03102802
CAS Number: 64096-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty