Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Methylpiperazin-1-yl)pyridin-2-amine 98%

Supplier: AMBEED, INC
Warning

A120592-10G A120592-1G A120592-25G
76781-730EA 43 USD InStock
76781-730 76781-732 76781-734
5-(4-Methylpiperazin-1-yl)pyridin-2-amine 98%
5-(4-Methylpiperazin-1-yl)pyridin-2-amine
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₄
MDL Number: MFCD10009563
CAS Number: 571189-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty