Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-iodonitrobenzene 96%

Supplier: AMBEED, INC
Warning

A123574-100G A123574-10G A123574-1G A123574-250MG A123574-25G A123574-5G
76781-852EA 123 USD InStock
76781-852 76781-854 76781-856 76781-858 76781-860 76781-862
2-Fluoro-5-iodonitrobenzene 96%
2-Fluoro-5-iodonitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃FINO₂
MW: 266.996 g/mol
MDL Number: MFCD09800783
CAS Number: 364-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty