Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-methoxypyrazine 98%

Supplier: AMBEED, INC
Warning

A125162-10G A125162-250MG A125162-25G A125162-5G
76781-946EA 22 USD InStock
76781-946 76781-948 76781-950 76781-952
2-Amino-5-bromo-3-methoxypyrazine 98%
2-Amino-5-bromo-3-methoxypyrazine
Precaución: for Research Use Only


Formula: C₅H₆BrN₃O
MDL Number: MFCD08275726
CAS Number: 5900-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty