Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine 95%

Supplier: AMBEED, INC
Warning

A125639-10G A125639-1G A125639-250MG A125639-5G
76781-988EA 942 USD InStock
76781-988 76781-990 76781-992 76781-994
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine 95%
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
Precaución: for Research Use Only


Formula: C₇H₅BrN₄
MDL Number: MFCD09834926
CAS Number: 857641-46-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty