Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-methoxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A128689-1G A128689-250MG A128689-5G
76782-178EA 15 USD InStock
76782-178 76782-180 76782-182
2-Amino-6-methoxybenzonitrile 98%
2-Amino-6-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD09745275
CAS Number: 1591-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty