Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate 98%

Supplier: AMBEED, INC
Warning

A129281-100MG A129281-250MG A129281-5G
76782-222EA 106 USD InStock
76782-222 76782-224 76782-226
Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate 98%
Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₂
MDL Number: MFCD16660980
CAS Number: 773136-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty