Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-3-methylbenzonitrile 98%

Supplier: AMBEED, INC
Warning

A130122-10G A130122-1G A130122-250MG A130122-25G A130122-5G
76782-300EA 63 USD InStock
76782-300 76782-302 76782-304 76782-306 76782-308
2-Chloro-4-fluoro-3-methylbenzonitrile 98%
2-Chloro-4-fluoro-3-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅ClFN
MW: 169.5834 g/mol
MDL Number: MFCD13193544
CAS Number: 796600-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty