Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium difluorotriphenylsilicate 97%

Supplier: AMBEED, INC
Danger

A135549-1G A135549-250MG A135549-25G A135549-5G
76782-628EA 7 USD InStock
76782-628 76782-630 76782-632 76782-634
Tetrabutylammonium difluorotriphenylsilicate 97%
Tetrabutylammonium difluorotriphenylsilicate
Precaución: for Research Use Only


Formula: C₃₄H₅₁F₂NSi
MDL Number: MFCD00274218
CAS Number: 163931-61-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty