Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one 95%

Supplier: AMBEED, INC
Warning

A136284-100MG A136284-250MG A136284-1G
76782-674EA 31 USD InStock
76782-674 76782-676 77666-966
5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one 95%
5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one
Precaución: for Research Use Only


Formula: C₆H₄BrN₃O
MDL Number: MFCD09999236
CAS Number: 22276-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty