Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate 97%

Supplier: AMBEED, INC
Warning

A136591-100G A136591-10G A136591-1G A136591-25G A136591-5G
76782-688EA 95 USD InStock
76782-688 76782-690 76782-692 76782-694 76782-696
7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate 97%
7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₄
MDL Number: MFCD09833010
CAS Number: 179688-53-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty