Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Xanthohumol 99%

Supplier: AMBEED, INC
Warning

A139015-100MG A139015-10MG A139015-1G A139015-1MG A139015-250MG A139015-25MG A139015-5MG
76782-838EA 162 USD InStock
76782-838 76782-840 76782-842 76782-844 76782-846 76782-848 76782-850
Xanthohumol 99%
Xanthohumol
Precaución: for Research Use Only


Formula: C₂₁H₂₂O₅
MDL Number: MFCD00210576
CAS Number: 6754-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty