Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-(4-Fluorophenyl)-4'-propyl-1,1'-bi(cyclohexane) 97%

Supplier: AMBEED, INC
Warning

A140957-1G A140957-250MG A140957-5G
76782-908EA 11 USD InStock
76782-908 76782-910 76782-912
trans-4-(4-Fluorophenyl)-4'-propyl-1,1'-bi(cyclohexane) 97%
trans-4-(4-Fluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Precaución: for Research Use Only


Formula: C₂₁H₃₁F
MDL Number: MFCD09838996
CAS Number: 82832-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty