Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanobenzophenone 95%

Supplier: AMBEED, INC
Warning

A141149-100MG A141149-1G A141149-250MG A141149-5G
76782-928EA 13 USD InStock
76782-928 76782-930 76782-932 76782-934
4-Cyanobenzophenone 95%
4-Cyanobenzophenone
Precaución: for Research Use Only


Formula: C₁₄H₉NO
MW: 207.23
Boiling Pt: 363
MDL Number: MFCD00016382
CAS Number: 1503-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty