Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-chlorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A141340-1G A141340-10G
76782-940EA 9 USD InStock
76782-940 77667-044
3-Bromo-4-chlorobenzonitrile 98%
3-Bromo-4-chlorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃BrClN
MDL Number: MFCD09025661
CAS Number: 948549-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty