Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromo-4-tert-butylbenzene 95%

Supplier: AMBEED, INC
Warning

A142827-100G A142827-10G A142827-25G A142827-5G
76782-994EA 256 USD InStock
76782-994 76782-996 76782-998 76783-000
1,2-Dibromo-4-tert-butylbenzene 95%
1,2-Dibromo-4-tert-butylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂Br₂
MDL Number: MFCD05664303
CAS Number: 6683-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty