Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-cyanothiazole 95%

Supplier: AMBEED, INC
Warning

A143404-100MG A143404-1G A143404-250MG
76783-020EA 12 USD InStock
76783-020 76783-022 76783-024
2-Bromo-4-cyanothiazole 95%
2-Bromo-4-cyanothiazole
Precaución: for Research Use Only


Formula: C₄HBrN₂S
MDL Number: MFCD09909650
CAS Number: 848501-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty