Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[2,1-b]thiazole-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A144146-100MG A144146-1G A144146-250MG
76783-074EA 45 USD InStock
76783-074 76783-076 76783-078
Imidazo[2,1-b]thiazole-2-carboxylic acid 97%
Imidazo[2,1-b]thiazole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₄N₂O₂S
MDL Number: MFCD13179636
CAS Number: 773841-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty