Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1,3-di-tert-butyl-2-methoxybenzene 98%

Supplier: AMBEED, INC
Warning

A145128-10G A145128-1G A145128-250MG A145128-5G A145128-25G
76783-114EA 57 USD InStock
76783-114 76783-116 76783-118 76783-120 77549-760
5-Bromo-1,3-di-tert-butyl-2-methoxybenzene 98%
5-Bromo-1,3-di-tert-butyl-2-methoxybenzene
Precaución: for Research Use Only


Formula: C₁₅H₂₃BrO
MDL Number: MFCD12024327
CAS Number: 1516-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty