Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Phenoxyphenyl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A146570-1G A146570-5G A146570-250MG
76783-182EA 16 USD InStock
76783-182 76783-184 77667-092
(3-Phenoxyphenyl)acetic acid 95%
(3-Phenoxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₅OC₆H₅)CH₂COOH
MDL Number: MFCD00016826
CAS Number: 32852-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty