Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-D-Val-OSu 97%

Supplier: AMBEED, INC
Warning

A146651-10G A146651-1G A146651-25G A146651-5G
76783-196EA 35 USD InStock
76783-196 76783-198 76783-200 76783-202
Z-D-Val-OSu 97%
Z-D-Val-OSu
Precaución: for Research Use Only


Formula: C₁₇H₂₀N₂O₆
MW: 348.355 g/mol
MDL Number: MFCD00153351
CAS Number: 4467-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty