Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-(trifluoromethyl)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A147415-100MG A147415-1G A147415-250MG A147415-5G
76783-226EA 8 USD InStock
76783-226 76783-228 76783-230 76783-232
2-Hydroxy-5-(trifluoromethyl)benzonitrile 95%
2-Hydroxy-5-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₄F₃NO
MW: 187.1187 g/mol
MDL Number: MFCD11213679
CAS Number: 142167-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty