Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BML-284 98+%

Supplier: AMBEED, INC
Warning

A149492-10MG A149492-1MG A149492-25MG A149492-5MG
76783-304EA 47 USD InStock
76783-304 76783-306 76783-308 76783-310
BML-284 98+%
BML-284
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₄O₃
MDL Number: MFCD09908087
CAS Number: 853220-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty