Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl N-(3-Trifluoromethylphenyl)anthranilate 98%

Supplier: AMBEED, INC
Warning

A149524-25G A149524-50MG A149524-5G
76783-312EA 34 USD InStock
76783-312 76783-314 76783-316
Butyl N-(3-Trifluoromethylphenyl)anthranilate 98%
Butyl N-(3-Trifluoromethylphenyl)anthranilate
Precaución: for Research Use Only


Formula: C₁₈H₁₈F₃NO₂
MW: 337.34 g/mol
MDL Number: MFCD00866001
CAS Number: 67330-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty