Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans,trans-4-Butyl-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane) 98%

Supplier: AMBEED, INC
Warning

A153758-100G A153758-10G A153758-25G A153758-5G
76783-498EA 56 USD InStock
76783-498 76783-500 76783-502 76783-504
trans,trans-4-Butyl-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane) 98%
trans,trans-4-Butyl-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
Precaución: for Research Use Only


Formula: C₂₂H₃₂F₂
MDL Number: MFCD11053391
CAS Number: 82832-58-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty