Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RepSox 98%

Supplier: AMBEED, INC
Warning

A158647-1MG A158647-50MG A158647-250MG A158647-100MG A158647-1G
76783-728EA 7 USD InStock
76783-728 77600-258 77667-234 77667-230 77667-232
RepSox 98%
RepSox
Precaución: for Research Use Only


Formula: C₁₇H₁₃N₅
MDL Number: MFCD09037561
CAS Number: 446859-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty