Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3,5-difluoro-2-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A158835-100G A158835-10G A158835-1G A158835-25G A158835-5G
76783-746EA 111 USD InStock
76783-746 76783-748 76783-750 76783-752 76783-754
1-Bromo-3,5-difluoro-2-nitrobenzene 98%
1-Bromo-3,5-difluoro-2-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD04112501
CAS Number: 884494-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty