Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-(Trifluoromethyl)biphenyl-3-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A160396-100MG A160396-5G
76783-792EA 36 USD InStock
76783-792 76783-794
2'-(Trifluoromethyl)biphenyl-3-carbaldehyde 97%
2'-(Trifluoromethyl)biphenyl-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₉F₃O
MDL Number: MFCD03424642
CAS Number: 675596-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty