Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1-(2-methylpropyl)-1H-imidazo[4,5-c]quinoline 97%

Supplier: AMBEED, INC
Warning

A170305-10G A170305-1G A170305-250MG A170305-25G A170305-5G
76784-190EA 29 USD InStock
76784-190 76784-192 76784-194 76784-196 76784-198
4-Chloro-1-(2-methylpropyl)-1H-imidazo[4,5-c]quinoline 97%
4-Chloro-1-(2-methylpropyl)-1H-imidazo[4,5-c]quinoline
Precaución: for Research Use Only


Formula: C₁₄H₁₄ClN₃
MDL Number: MFCD06658174
CAS Number: 99010-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty